Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046049.2 | NP_001039514.1 | 8 - 9 | 1147 | 73 | |
| GenBank mRNA | BC105478.1 | - | 8 - 9 | 1148 | 73 | |
More Information
Set Membership: |
|

VPS33A
Genomic Map