Assay Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046086.2 | NP_001039551.1 | 3 - 4 | 413 | 74 | |
| GenBank mRNA | BC112552.1 | - | 3 - 4 | 414 | 74 | |
More Information
Set Membership: |
|

WDR54
Genomic Map