Assay Details
| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046128.1 | NP_001039593.1 | 2 - 3 | 219 | 78 | |
| GenBank mRNA | BC105568.1 | - | 2 - 3 | 219 | 78 | |
| BT030706.1 | - | 2 - 3 | 184 | 78 | ||
More Information
Set Membership: |
|

PHKG2
Genomic Map