Assay Details
| Gene Symbol: GFUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046139.3 | NP_001039604.1 | 9 - 10 | 909 | 69 | |
| GenBank mRNA | BC112514.1 | - | 9 - 10 | 889 | 69 | |
More Information
Set Membership: |
|

GFUS
Genomic Map