Assay Details
| Gene Symbol: PPP1R15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046178.2 | NP_001039643.1 | 1 - 2 | 222 | 133 | |
| GenBank mRNA | BC112768.1 | - | 1 - 2 | 223 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112768.1, NM_001046178.2 transcripts.
Set Membership: |
|

PPP1R15A
Genomic Map