Assay Details
| Gene Symbol: ISG20L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046217.2 | NP_001039682.1 | 2 - 3 | 870 | 64 | |
| GenBank mRNA | BC110186.1 | - | 2 - 3 | 871 | 64 | |
More Information
Set Membership: |
|

ISG20L2
Genomic Map