Assay Details
| Gene Symbol: ISG20L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046217.2 | NP_001039682.1 | 3 - 4 | 1073 | 77 | |
| GenBank mRNA | BC110186.1 | - | 3 - 4 | 1074 | 77 | |
More Information
Set Membership: |
|

ISG20L2
Genomic Map