Assay Details
| Gene Symbol: CDRT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046220.2 | NP_001039685.1 | 3 - 4 | 307 | 64 | |
| GenBank mRNA | BC110162.1 | - | 3 - 4 | 308 | 64 | |
More Information
Set Membership: |
|

CDRT4
Genomic Map