Assay Details
| Gene Symbol: LOC518526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046239.2 | NP_001039704.2 | 4 - 5 | 485 | 105 | |
| NM_001278620.1 | NP_001265549.1 | 3 - 4 | 362 | 105 | ||
| GenBank mRNA | BC112602.1 | - | 4 - 5 | 485 | 105 | |
| DV808040.1 | - | 2 - 3 | 329 | 105 | ||
More Information
Set Membership: |
|

LOC518526
Genomic Map