Assay Details
| Gene Symbol: H2BC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046246.2 | NP_001039711.1 | 1 - 2 | 472 | 74 | |
| GenBank mRNA | BC112625.1 | - | 1 - 2 | 473 | 74 | |
More Information
Set Membership: |
|

H2BC5
Genomic Map