Assay Details
| Gene Symbol: CALM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046249.2 | NP_001039714.1 | 4 - 5 | 373 | 54 | |
| GenBank mRNA | BC105380.1 | - | 4 - 5 | 374 | 54 | |
More Information
Set Membership: |
|

CALM3
Genomic Map