Assay Details
| Gene Symbol: NEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046270.2 | NP_001039735.1 | 4 - 5 | 738 | 72 | |
| GenBank mRNA | BC112555.1 | - | 4 - 5 | 739 | 72 | |
More Information
Set Membership: |
|

NEK2
Genomic Map