Assay Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046294.1 | NP_001039759.1 | 5 - 6 | 689 | 64 | |
| GenBank mRNA | BC112528.1 | - | 5 - 6 | 689 | 64 | |
More Information
Set Membership: |
|

DHRS1
Genomic Map