Assay Details
| Gene Symbol: TDO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046313.1 | NP_001039778.1 | 2 - 3 | 197 | 72 | |
| GenBank mRNA | BC112550.1 | - | 2 - 3 | 197 | 72 | |
More Information
Set Membership: |
|

TDO2
Genomic Map