Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046332.1 | NP_001039797.1 | 3 - 4 | 247 | 67 | |
| GenBank mRNA | BC105461.1 | - | 3 - 4 | 247 | 67 | |
| FJ358601.1 | - | 2 - 3 | 244 | 67 | ||
More Information
Set Membership: |
|

CDC42
Genomic Map