Assay Details
| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046347.2 | NP_001039812.1 | 8 - 9 | 1265 | 70 | |
| GenBank mRNA | BC112787.1 | - | 8 - 9 | 1267 | 70 | |
More Information
Set Membership: |
|

RAB3IP
Genomic Map