Assay Details
| Gene Symbol: STING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046357.2 | NP_001039822.1 | 3 - 4 | 377 | 83 | |
| GenBank mRNA | BC112716.1 | - | 3 - 4 | 378 | 83 | |
| KU695665.1 | - | 2 - 3 | 306 | 83 | ||
| KU695666.1 | - | 2 - 3 | 306 | 83 | ||
More Information
Set Membership: |
|

STING1
Genomic Map