Assay Details
| Gene Symbol: STING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046357.2 | NP_001039822.1 | 6 - 7 | 915 | 70 | |
| GenBank mRNA | BC112716.1 | - | 6 - 7 | 916 | 70 | |
| KU695665.1 | - | 5 - 6 | 844 | 70 | ||
More Information
Set Membership: |
|

STING1
Genomic Map