Assay Details
| Gene Symbol: FKBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046372.2 | NP_001039837.1 | 1 - 2 | 358 | 62 | |
| GenBank mRNA | BC105407.1 | - | 1 - 2 | 360 | 62 | |
More Information
Set Membership: |
|

FKBP9
Genomic Map