Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046377.1 | NP_001039842.1 | 1 - 2 | 299 | 147 | |
| GenBank mRNA | BC105474.1 | - | 1 - 2 | 299 | 147 | |
| BT030528.1 | - | 1 - 2 | 328 | 147 | ||
More Information
Set Membership: |
|

TMED5
Genomic Map