Assay Details
| Gene Symbol: CWC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046399.2 | NP_001039864.1 | 5 - 6 | 605 | 134 | |
| GenBank mRNA | BC105398.1 | - | 5 - 6 | 606 | 134 | |
More Information
Set Membership: |
|

CWC15
Genomic Map