Assay Details
| Gene Symbol: FAM162A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046592.1 | NP_001040057.1 | 2 - 3 | 224 | 65 | |
| GenBank mRNA | BC111680.1 | - | 2 - 3 | 224 | 65 | |
More Information
Set Membership: |
|

FAM162A
Genomic Map