Assay Details
| Gene Symbol: CCNDBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046604.1 | NP_001040069.1 | 6 - 7 | 591 | 60 | |
| GenBank mRNA | BC112447.1 | - | 6 - 7 | 591 | 60 | |
More Information
Set Membership: |
|

CCNDBP1
Genomic Map