Assay Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046609.2 | NP_001040074.1 | 2 - 3 | 455 | 60 | |
| GenBank mRNA | BC110133.1 | - | 2 - 3 | 456 | 60 | |
More Information
Set Membership: |
|

TSKS
Genomic Map