Assay Details
| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075292.2 | NP_001068760.1 | 2 - 3 | 409 | 61 | |
| GenBank mRNA | BC111652.1 | - | 2 - 3 | 410 | 61 | |
More Information
Set Membership: |
|

SEC61B
Genomic Map