Assay Details
| Gene Symbol: MRPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075619.2 | NP_001069087.1 | 1 - 2 | 173 | 59 | |
| GenBank mRNA | BC110245.1 | - | 1 - 2 | 174 | 59 | |
More Information
Set Membership: |
|

MRPS35
Genomic Map