Assay Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075928.2 | NP_001069396.1 | 1 - 2 | 95 | 74 | |
| GenBank mRNA | BC110149.1 | - | 1 - 2 | 96 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110149.1, NM_001075928.2 transcripts.
Set Membership: |
|

GLT6D1
Genomic Map