Assay Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075928.2 | NP_001069396.1 | 4 - 5 | 355 | 114 | |
| GenBank mRNA | BC110149.1 | - | 4 - 5 | 356 | 114 | |
More Information
Set Membership: |
|

GLT6D1
Genomic Map