Assay Details
| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077068.1 | NP_001070536.1 | 5 - 6 | 572 | 69 | |
| GenBank mRNA | BC110220.1 | - | 5 - 6 | 572 | 69 | |
More Information
Set Membership: |
|

TMBIM7
Genomic Map