Assay Details
| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077069.1 | NP_001070537.2 | 1 - 2 | 172 | 83 | |
| GenBank mRNA | BC110240.1 | - | 1 - 2 | 172 | 83 | |
More Information
Set Membership: |
|

TMEM247
Genomic Map