Assay Details
| Gene Symbol: KRTAP11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080740.1 | NP_001074209.1 | 1 | 755 | 77 | |
| GenBank mRNA | BC122847.1 | - | 1 | 755 | 77 | |
More Information
Set Membership: |
|

KRTAP11-1
Genomic Map