Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080918.1 | NP_001074387.1 | 2 - 3 | 249 | 110 | |
| GenBank mRNA | BC110007.1 | - | 2 - 3 | 249 | 110 | |
More Information
Set Membership: |
|

DNAL4
Genomic Map