Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011676.3 | NP_001011676.2 | 3 - 4 | 626 | 70 | |
| GenBank mRNA | BC118117.1 | - | 3 - 4 | 627 | 70 | |
More Information
Set Membership: |
|

CD2
Genomic Map