Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014870.3 | NP_001014870.1 | 8 - 9 | 806 | 62 | |
| GenBank mRNA | BC114088.1 | - | 8 - 9 | 807 | 62 | |
| BT020679.1 | - | 8 - 9 | 798 | 62 | ||
More Information
Set Membership: |
|

TMEM106A
Genomic Map