Assay Details
| Gene Symbol: MYL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015640.2 | NP_001015640.1 | 2 - 3 | 200 | 65 | |
| GenBank mRNA | BC118244.1 | - | 2 - 3 | 200 | 65 | |
| BT020978.1 | - | 2 - 3 | 200 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118244.1, NM_001015640.2 transcripts.
Set Membership: |
|

MYL12A
Genomic Map