Assay Details
| Gene Symbol: FOXA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033119.2 | NP_001028291.1 | 1 - 2 | 75 | 71 | |
| GenBank mRNA | BC116076.1 | - | 1 - 2 | 75 | 71 | |
| DQ157763.1 | - | 1 - 2 | 67 | 71 | ||
More Information
Set Membership: |
|

FOXA3
Genomic Map