Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045867.1 | NP_001039332.1 | 6 - 7 | 937 | 84 | |
| GenBank mRNA | BC114101.1 | - | 6 - 7 | 937 | 84 | |
More Information
Set Membership: |
|

GYG1
Genomic Map