Assay Details
| Gene Symbol: CHORDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045912.2 | NP_001039377.1 | 8 - 9 | 765 | 80 | |
| GenBank mRNA | BC114125.1 | - | 8 - 9 | 766 | 80 | |
More Information
Set Membership: |
|

CHORDC1
Genomic Map