Assay Details
| Gene Symbol: MINDY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045924.1 | NP_001039389.1 | 5 - 6 | 1185 | 57 | |
| GenBank mRNA | BC105561.1 | - | 5 - 6 | 1185 | 57 | |
More Information
Set Membership: |
|

MINDY1
Genomic Map