Assay Details
| Gene Symbol: TCEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045925.1 | NP_001039390.1 | 4 - 5 | 463 | 63 | |
| GenBank mRNA | BC114117.1 | - | 4 - 5 | 463 | 63 | |
More Information
Set Membership: |
|

TCEA1
Genomic Map