Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045943.2 | NP_001039408.1 | 8 - 9 | 793 | 118 | |
| NM_001142365.1 | NP_001135837.1 | 9 - 10 | 907 | 118 | ||
| GenBank mRNA | BC112581.1 | - | 8 - 9 | 767 | 118 | |
More Information
Set Membership: |
|

ALG5
Genomic Map