Assay Details
| Gene Symbol: SPINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045971.1 | NP_001039436.1 | 1 - 2 | 437 | 64 | |
| GenBank mRNA | BC105512.1 | - | 1 - 2 | 437 | 64 | |
More Information
Set Membership: |
|

SPINT2
Genomic Map