Assay Details
| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045997.2 | NP_001039462.1 | 8 - 9 | 817 | 65 | |
| GenBank mRNA | BC112649.1 | - | 8 - 9 | 818 | 65 | |
More Information
Set Membership: |
|

CPB2
Genomic Map