Assay Details
| Gene Symbol: MRPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046068.2 | NP_001039533.1 | 2 - 3 | 243 | 97 | |
| GenBank mRNA | BC105368.1 | - | 2 - 3 | 244 | 97 | |
More Information
Set Membership: |
|

MRPL19
Genomic Map