Assay Details
| Gene Symbol: LY6E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046070.1 | NP_001039535.1 | 3 - 4 | 358 | 64 | |
| GenBank mRNA | BC114823.1 | - | 3 - 4 | 358 | 64 | |
More Information
Set Membership: |
|

LY6E
Genomic Map