Assay Details
| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046095.2 | NP_001039560.1 | 1 - 2 | 87 | 83 | |
| GenBank mRNA | BC114022.1 | - | 1 - 2 | 88 | 83 | |
| JC209428.1 | - | 1 - 2 | 88 | 83 | ||
More Information
Set Membership: |
|

CXCL16
Genomic Map