Assay Details
| Gene Symbol: TVP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046152.2 | NP_001039617.1 | 5 - 6 | 576 | 61 | |
| GenBank mRNA | BC113231.1 | - | 5 - 6 | 472 | 61 | |
| EH153594.1 | - | 5 - 6 | 576 | 61 | ||
More Information
Set Membership: |
|

TVP23B
Genomic Map