Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046176.2 | NP_001039641.1 | 3 - 4 | 302 | 116 | |
| GenBank mRNA | BC105431.1 | - | 3 - 4 | 303 | 116 | |
More Information
Set Membership: |
|

SEPTIN10
Genomic Map