Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046216.1 | NP_001039681.1 | 7 - 8 | 1533 | 67 | |
| GenBank mRNA | BC113317.1 | - | 7 - 8 | 1533 | 67 | |
More Information
Set Membership: |
|

RNF8
Genomic Map