Assay Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046224.1 | NP_001039689.1 | 5 - 6 | 851 | 77 | |
| GenBank mRNA | BC114087.1 | - | 5 - 6 | 851 | 77 | |
More Information
Set Membership: |
|

USP1
Genomic Map